Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perfluorooctyl bromide, 99%

Catalog No. AC386790500
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle

CAS: 423-55-2 | C8BrF17 | 498.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 423-55-2
Molecular Formula C8BrF17
Molecular Weight (g/mol) 498.97
MDL Number MFCD00042082
InChI Key WTWWXOGTJWMJHI-UHFFFAOYSA-N
Synonym perflubron, 1-bromoperfluorooctane, perfluorooctyl bromide, 1-bromoheptadecafluorooctane, oxygent, perfluoro-n-octyl bromide, imagent gi, perfluorooctylbromide, liquivent, perfluoroctyl bromide
PubChem CID 9873
ChEBI CHEBI:38803
IUPAC Name 1-bromo-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluorooctane
SMILES FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)Br

Specifications

Melting Point 6.0°C
Color Colorless to Tan
Density 1.9300g/mL
Boiling Point 142.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.3050 to 1.3070
Linear Formula CF3(CF2)7Br
Quantity 50 g
Merck Index 15,727
Specific Gravity 1.93
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with organic solvents
Viscosity 2.3 mPa.s (20°C)
Formula Weight 498.95
Percent Purity 99%
Physical Form Viscous Solution
Chemical Name or Material Perfluorooctyl bromide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 207-028-4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.