Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Serine methyleester hydrochloride, 98%, Thermo Scientific Chemicals

Catalog No. AC396790250
Click to view available options
Quantity:
25 g
Packaging:
Glass Bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5874-57-7
Molecular Formula C23H19Br2N3O3S
Molecular Weight (g/mol) 577.29
MDL Number MFCD00066121
InChI Key LYRWWUCOZBTOPL-UHFFFAOYNA-N
Synonym d-serine methyl ester hydrochloride, h-d-ser-ome.hcl, r-methyl 2-amino-3-hydroxypropanoate hydrochloride, d-serine, methyl ester, hydrochloride, d-serine methyl ester hcl, methyl 2r-2-amino-3-hydroxypropanoate hydrochloride, d-ser-ome hcl, cycloserine impurity 2, h-d-ser-ome.hci
PubChem CID 11446470
IUPAC Name methyl (2R)-2-amino-3-hydroxypropanoate;hydrochloride
SMILES CC(N(C)S(=O)(=O)C1=CC=C(Br)C=C1)C1=NC2=CC=CC=C2C(=O)N1C1=CC=C(Br)C=C1

Specifications

Melting Point 152.0°C to 161.0°C
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula HOCH2CH(NH2)CO2CH3·HCl
Specific Rotation −2.5° to −4.5° (20°C, 589nm) (C=4, methanol)
Quantity 25 g
Formula Weight 155.58
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material D-Serine methyl ester hydrochloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.