Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perforin Antibody (CE2.10) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$268.00 - $608.50

Specifications

Antigen Perforin
Clone CE2.10
Concentration 1.0 mg/mL
Dilution Western Blot 1:100-1:2000, Immunoprecipitation 1:10-1:500
Applications Western Blot, Immunoprecipitation
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB018763
SDP
View Documents
Novus Biologicals
NBP1974400.025MG
0.025 mg
Each for $268.00
Only null left
Add to Cart
 
NBP197440
SDP
View Documents
Novus Biologicals
NBP197440
0.1 mg
Each for $608.50
Only null left
Add to Cart
 
Description

Description

Perforin Monoclonal specifically detects Perforin in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

Perforin
1.0 mg/mL
Western Blot, Immunoprecipitation
Unconjugated
Mouse
Innate Immunity
Cytolysin, FLH2, HPLH2, HPLH2lymphocyte pore forming protein, Lymphocyte pore-forming protein, MGC65093, P1PFN1, perforin 1 (pore forming protein), perforin-1, PFPcytolysin
PRF1
IgG2a
Protein A or G purified
Recognizes an epitope in the region aa 402-534 of human and mouse perforin. Does NOT detect Human in Western Blot.
CE2.10
Western Blot 1:100-1:2000, Immunoprecipitation 1:10-1:500
Monoclonal
Purified
RUO
Human, Mouse
5551
Recombinant mouse perforin (aa 98-534).
Primary
Store at 4C. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.