Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Perforin Antibody (CE2.10), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP197440V
Description
Perforin Monoclonal specifically detects Perforin in Human, Mouse samples. It is validated for Western Blot.Specifications
Perforin | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PRF1 | |
Recombinant mouse perforin (aa 98-534). | |
0.1 mL | |
Adaptive Immunity, Apoptosis, Immunology, Innate Immunity | |
Recognizes an epitope in the region aa 402-534 of human and mouse perforin. Does NOT detect Human in Western Blot. | |
Store at 4°C in the dark. |
Western Blot | |
CE2.10 | |
Western Blot | |
Cytolysin, FLH2, HPLH2, HPLH2lymphocyte pore forming protein, Lymphocyte pore-forming protein, MGC65093, P1PFN1, perforin 1 (pore forming protein), perforin-1, PFPcytolysin | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction