Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Perforin Antibody (PRF1/2470) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP279886
Description
Perforin Monoclonal specifically detects Perforin in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
Perforin | |
Monoclonal | |
1 mg/mL | |
ELISA, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
Cytolysin, FLH2, HPLH2, HPLH2lymphocyte pore forming protein, Lymphocyte pore-forming protein, MGC65093, P1PFN1, perforin 1 (pore forming protein), perforin-1, PFPcytolysin | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
IgG2c κ |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
PRF1/2470 | |
Unconjugated | |
10 mM PBS with No Preservative | |
PRF1 | |
Recombinant human Perforin protein fragment (around aa 413-552) (exact sequence is proprietary) (Uniprot: P14222) | |
100 μg | |
Adaptive Immunity, Apoptosis, Immunology, Innate Immunity | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction