Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Perforin Monoclonal Antibody (CB5.4)
GREENER_CHOICE

Catalog No. PIMA125830
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA125830 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA125830 Supplier Invitrogen™ Supplier No. MA125830
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Positive control of mouse T cell clone B6.1 suggested.

Perforin is one of the major cytolytic proteins of cytolytic granules. Perforin is a cytolytic mediator and is stored in and released by cytoplasmic granules. Moreover, perforin is involved in immune defense against tumors and virus infections as mediated by cytotoxic lymphocytes. Perforin is a 555 amino acid protein with a 21 amino acid signal peptide, and has a molecular weight of 70 to 75 kD. Perforin is a pore forming protein with a mechanism of transmembrane channel formation similar to C9, and homology between perforin and C9 have been demonstrated. Studies show that perforin is expressed only in killer cell lines and not in helper T lymphocytes or other tumor cells tested. Perforin is known to be a key effector molecule for T-cell- and natural killer-cell-mediated cytolysis. Defects in the perforin gene cause familial hemophagocytic lymphohistiocytosis type 2 (HPLH2), a rare and lethal autosomal recessive disorder of early childhood. Alternative splicing results in multiple transcript variants of perforin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Perforin
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone CB5.4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene PRF1
Gene Accession No. P10820
Gene Alias Cyta; Cytolysin; FLH2; HPLH2; lymphocyte pore forming protein; lymphocyte pore-forming protein; OMAK; OTTHUMP00000019759; P1; perforin 1; perforin 1 (pore forming protein); Perforin1; perforin-1; Pfn; PFN1; Pfp; PGFL; PIGF; PIGF-2; PLGF; pore forming protein; pore-forming protein; PRF1; Prf-1; PRF1 (pore forming protein 1); RATCYTA; RP11-710A11.3
Gene Symbols PRF1
Host Species Rat
Immunogen Recombinant mouse perforin, amino acids 98-534.
Purification Method IgG fraction
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18646
Target Species Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.