Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HexArmor™ 9000 Series™ 9011 Palm-Coated Gloves

Catalog No. 19160620 Shop All Performance Fabrics, Inc Products
Encompass
Click to view available options
Size:
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Lightweight, cut-resistant gloves are constructed of a red cotton shell, rubber palm coating and interior layer of SuperFabric™ brand material for comfort and safety.

  • Made with SuperFabric™ brand material with ANSI/ISEA Level A7 cut protection
  • Industrial puncture protection on enhanced areas from wires, metal, wood and glass
  • Comfortable cotton blend shell

Specifications

Coating Material Rubber
Color Red, Black
Cut Resistance Rating ANSI A7
Material SuperFabric, Cotton Blend
Product Type Cut-resistant Gloves
Size X-Large
EN388 Standard EN 388:4X44F
For Use With (Application) General work, waste handling, maintenance, glass, metal and wire handling
Model 9011-10
Series 9000 Series
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.