Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Performance Plastic Chemical Wash Bottles with 2 Color Labels
SCI_ED

Catalog No. S27151 Shop All Eisco Products
Change view
Click to view available options
Capacity (Metric):
500 mL
1000 mL
Chemical Label:
Acetone
Distilled Water
Ethanol
Isopropanol
Methanol
Sodium Hypochlorite (Bleach)
Quantity:
1/Pk.
2/Pk.
6/Pk.
32 product options available for selection
Product selection table with 32 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Chemical Label Quantity
S27151 500 mL Acetone 1/Pk.
S27152 500 mL Acetone 2/Pk.
S27153 500 mL Acetone 6/Pk.
S27160 1000 mL Acetone 1/Pk.
S27161 1000 mL Acetone 2/Pk.
S27162 1000 mL Acetone 6/Pk.
S27169 500 mL Distilled Water 1/Pk.
S27170 500 mL Distilled Water 2/Pk.
S27171 500 mL Distilled Water 6/Pk.
S27178 1000 mL Distilled Water 1/Pk.
S27179 1000 mL Distilled Water 2/Pk.
S27180 1000 mL Distilled Water 6/Pk.
S27187 500 mL Isopropanol 1/Pk.
S27188 500 mL Isopropanol 2/Pk.
S27189 500 mL Isopropanol 6/Pk.
S27196 1000 mL Isopropanol 1/Pk.
S27197 1000 mL Isopropanol 2/Pk.
S27198 1000 mL Isopropanol 6/Pk.
S27205 500 mL Ethanol 1/Pk.
S27206 500 mL Ethanol 2/Pk.
S27207 500 mL Ethanol 6/Pk.
S27214 1000 mL Ethanol 1/Pk.
S27215 1000 mL Ethanol 2/Pk.
S27216 1000 mL Ethanol 6/Pk.
S27223 500 mL Sodium Hypochlorite (Bleach) 1/Pk.
S27224 500 mL Sodium Hypochlorite (Bleach) 2/Pk.
S27225 500 mL Sodium Hypochlorite (Bleach) 6/Pk.
S27232 1000 mL Sodium Hypochlorite (Bleach) 1/Pk.
S27233 1000 mL Sodium Hypochlorite (Bleach) 2/Pk.
S27234 1000 mL Sodium Hypochlorite (Bleach) 6/Pk.
S27238 500 mL Methanol 1/Pk.
S27240 1000 mL Methanol 6/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 32 options available.
32 options
Catalog No. S27151 Supplier Eisco™ Supplier No. CHWB1011
Add to Cart
Add to Cart

Performance Plastic Wash Bottles are made of low-density polyethylene (LDPE) for superior quality, strength, and flexibility. Chemical labels feature the chemical information as well as hazard, safety, and first aid precautionary statements.

  • Chemical specific wash bottles with color coded caps and labels
  • Performance plastic is durable yet extremely responsive
  • Made of low-density polyethylene (LDPE)
  • Easy to read labeling, including chemical specific information and safety warnings
  • Self-venting design to eliminate leaks and dripping
  • Wide mouth for easy filling
  • Innovative flex ink won't fade, crack, or peel
  • Sodium Hypochlorite (bleach) wash bottles are opaque (white) to protect contents from UV light rays
  • Other bottles are clear/transparent
  • Bottle labels comply with the Globally Harmonized System of Classification and Labeling of Chemicals (GHS)
  • Bottle Labels also comply with OSHA's HazCom 2012 requirements and Hazard Communication Standard 29 CFR 1910.1200

Specifications

Capacity (Metric) 500 mL
Capacity (English) 17 oz.
Color Clear
Description Color coded
Diameter (English) 2.9 in.
Diameter (Metric) Inner 43.18 mm
Includes Built in nozzle
Height (Metric) With Closure 228.6 mm
Material LDPE
Length (Metric) 76.2 mm
No. per Pack 1
Quantity 1/Pk.
Sterility Non-sterile
Weight (Metric) 45.35 g
Weight (English) 0.1 lb.
Width (Metric) 76.2 mm
Closure Color Red
Closure Included Yes
Disposable Reusable
Graduated No
Mouth Narrow
Shape Cylindrical
Height (Metric) 133.35 mm
Product Type Wash Bottle
Width (English) 3 in.
Length (English) 3 in.
Height (English) 9 in.
Chemical Label Acetone
Closure Material Plastic
Closure Size 5 cm
Diameter (Metric) 73.66 mm
Closure Type Screw Cap
Autoclavable Not Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.