Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Performance Plastic Chemical Wash Bottles with 4 Color Labels
SCI_ED

Catalog No. S27154 Shop All Eisco Products
Change view
Click to view available options
Capacity (Metric):
500 mL
1000 mL
Chemical Label:
Acetone
Distilled Water
Ethanol
Isopropanol
Methanol
Sodium Hypochlorite (Bleach)
Quantity:
1/Pk.
2/Pk.
6/Pk.
32 product options available for selection
Product selection table with 32 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Chemical Label Quantity
S27154 500 mL Acetone 1/Pk.
S27155 500 mL Acetone 2/Pk.
S27156 500 mL Acetone 6/Pk.
S27163 1000 mL Acetone 1/Pk.
S27164 1000 mL Acetone 2/Pk.
S27165 1000 mL Acetone 6/Pk.
S27172 500 mL Distilled Water 1/Pk.
S27173 500 mL Distilled Water 2/Pk.
S27174 500 mL Distilled Water 6/Pk.
S27181 1000 mL Distilled Water 1/Pk.
S27182 1000 mL Distilled Water 2/Pk.
S27183 1000 mL Distilled Water 6/Pk.
S27190 500 mL Isopropanol 1/Pk.
S27191 500 mL Isopropanol 2/Pk.
S27192 500 mL Isopropanol 6/Pk.
S27199 1000 mL Isopropanol 1/Pk.
S27200 1000 mL Isopropanol 2/Pk.
S27201 1000 mL Isopropanol 6/Pk.
S27208 500 mL Ethanol 1/Pk.
S27209 500 mL Ethanol 2/Pk.
S27210 500 mL Ethanol 6/Pk.
S27217 1000 mL Ethanol 1/Pk.
S27218 1000 mL Ethanol 2/Pk.
S27219 1000 mL Ethanol 6/Pk.
S27226 500 mL Sodium Hypochlorite (Bleach) 1/Pk.
S27227 500 mL Sodium Hypochlorite (Bleach) 2/Pk.
S27228 500 mL Sodium Hypochlorite (Bleach) 6/Pk.
S27235 1000 mL Sodium Hypochlorite (Bleach) 1/Pk.
S27236 1000 mL Sodium Hypochlorite (Bleach) 2/Pk.
S27237 1000 mL Sodium Hypochlorite (Bleach) 6/Pk.
S27239 500 mL Methanol 1/Pk.
S27241 1000 mL Methanol 1/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 32 options available.
32 options
Catalog No. S27154 Supplier Eisco™ Supplier No. CHWB1012
Add to Cart
Add to Cart

Performance Plastic Wash Bottles are made of low-density polyethylene (LDPE) for superior quality, strength, and flexibility. Chemical labels feature the chemical information as well as hazard, safety, and first aid precautionary statements.

  • Chemical specific wash bottles with color coded caps and labels
  • Performance plastic is durable yet extremely responsive
  • Made of low-density polyethylene (LDPE)
  • Easy to read labeling, including chemical specific information and safety warnings
  • Self-venting design to eliminate leaks and dripping
  • Wide mouth for easy filling
  • Innovative flex ink won't fade, crack, or peel
  • Sodium Hypochlorite (bleach) wash bottles are opaque (white) to protect contents from UV light rays
  • Other bottles are clear/transparent
  • Bottle labels comply with the Globally Harmonized System of Classification and Labeling of Chemicals (GHS)
  • Bottle Labels also comply with OSHA's HazCom 2012 requirements and Hazard Communication Standard 29 CFR 1910.1200

Specifications

Capacity (Metric) 500 mL
Capacity (English) 17 oz.
Color Clear
Description Leak proof
Diameter (English) 2.9 in.
For Use With (Application) Used to rinse various pieces of laboratory glassware, such as test tubes and round bottom flasks.
Includes Built in nozzle
Material LDPE
Length (Metric) 76.2 mm
No. per Pack 1
Quantity 1/Pk.
Sterility Non-sterile
Width (Metric) 76.2 mm
Closure Color Red
Closure Included Yes
Disposable Reusable
Graduated No
Mouth Narrow
Shape Cylindrical
Height (Metric) 22.86 cm
Product Type Wash Bottle
For Use With (Equipment) Acetone
Width (English) 3 in.
Length (English) 3 in.
Height (English) 9 in.
Chemical Label Acetone
Closure Material Plastic
Closure Size 5 cm
Diameter (Metric) 7.36 cm
Closure Type Screw Cap
Autoclavable Not Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.