Learn More
Description
- Chemical specific wash bottles with color coded caps and labels
- Performance plastic is durable yet extremely responsive
- Made of low-density polyethylene (LDPE)
- Easy to read labeling, including chemical specific information and safety warnings
- Self-venting design to eliminate leaks and dripping
- Wide mouth for easy filling
- Innovative flex ink won't fade, crack, or peel
- Sodium Hypochlorite (bleach) wash bottles are opaque (white) to protect contents from UV light rays
- Other bottles are clear/transparent
- Bottle labels comply with the Globally Harmonized System of Classification and Labeling of Chemicals (GHS)
- Bottle Labels also comply with OSHA's HazCom 2012 requirements and Hazard Communication Standard 29 CFR 1910.1200
Specifications
Specifications
| Capacity (Metric) | 500 mL |
| Capacity (English) | 17 oz. |
| Color | Clear |
| Description | Color coded |
| For Use With (Application) | Used to rinse various pieces of laboratory glassware, such as test tubes and round bottom flasks. |
| Includes | Built in nozzle |
| Material | LDPE |
| No. per Pack | 2 |
| Quantity | 1/Pk. |
| Sterility | Non-sterile |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
