Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pergolide Mesylate, USP, 97.5-102%, Spectrum™ Chemical
SDP

Catalog No. 18605028
Click to view available options
Quantity:
100 mg
500 mg
1 g
5 g
25 g
100 g
Packaging:
Amber Glass Bottle

P3682, 66104-23-2, C19H26N2S·CH4O3S

Spectrum™ Chemical Pergolide Mesylate, USP is indicated as adjunctive treatment to levodopa/carbidopa in the management of the signs and symptoms of Parkinson†s disease. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 66104-23-2
Molecular Formula C20H30N2O3S2
Molecular Weight (g/mol) 410.59
InChI Key UWCVGPLTGZWHGS-ZORIOUSZSA-N
IUPAC Name (2R,4R,7R)-4-[(methylsulfanyl)methyl]-6-propyl-6,11-diazatetracyclo[7.6.1.0²,⁷.0¹²,¹⁶]hexadeca-1(16),9,12,14-tetraene; methanesulfonic acid
SMILES CS(O)(=O)=O.CCCN1C[C@H](CSC)C[C@H]2[C@H]1CC1=CNC3=CC=CC2=C13

Specifications

CAS Max % 1
Ignition Residue 0.001
Loss on Drying 0.005
Packaging Amber Glass Bottle
Quantity 500 mg
Specific Rotation -17° to -23°
Percent Purity 97.5 to 102%
Grade USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.