Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pericentrin Antibody, Alexa Fluor™ 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10061071AF594
Description
Pericentrin Polyclonal specifically detects Pericentrin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Pericentrin | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
Kendrin, KIAA0402, MOPD2, PCN, PCNT2pericentrin B, PCNTB, pericentrin, pericentrin 2 (kendrin), pericentrin, kendrin (KIAA0402)10KENPCTN2, pericentrin-2, pericentrin-380, pericentrin-B, SCKL4kendrin | |
Rabbit | |
100 μL | |
5116 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
PCNT | |
The immunogen recognized by this antibody maps to a region between residue 175 and 225 of human Pericentrin/Kendrin using the numbering given in entry NP_006022.3 (GeneID 5116). | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction