Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pericentrin/Kendrin Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501571786
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-1786 10 μL
50-157-1785 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-157-1786 Supplier Bethyl Laboratories, Inc Supplier No. IHC00264T
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Pericentrin 2, also known as Pericentrin B or Kendrin, is an integral component of the pericentriolar material. The protein localizes specifically to centrosomes throughout all stages of the cell cycle. The protein remains centrosomal following microtubule depolymerization. Pericentrin 2 binds calmodulin and is widely expressed in most tissues, including kidney, placenta, liver and thymus.

Specifications

Antigen Pericentrin/Kendrin
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene PCNT
Gene Accession No. O95613
Gene Alias Pericentrin, Kendrin, Pericentrin-B, PCNT, KIAA0402, PCNT2
Gene Symbols PCNT
Host Species Rabbit
Immunogen Between 175 and 225
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5116
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.