Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Pericentrin Recombinant Rabbit Monoclonal Antibody (PSH0-66)
GREENER_CHOICE

Catalog No. MA551929
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA551929 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA551929 Supplier Invitrogen™ Supplier No. MA551929
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Sequence Similarities: 63% Mouse/No Cross with Rat. Tissue Specificity: Expressed in all tissues tested, including placenta, liver, kidney and thymus. Positive Control: HeLa cell lysate, HepG2 cell lysate, MCF7 cell lysate, K-562 cell lysate. Subcellular Location: Cytoplasm, cytoskeleton, microtubule organizing center, centrosome. Predicted band size: 378 kDa.

This gene encodes a protein that was identified in a screen for genes expressed in metastatic cells, specifically, mammary adenocarcinoma cell lines. Expression of this gene has been correlated with the metastatic potential of at least two types of carcinomas although it is also expressed in many normal tissues. The role it plays in metastasis is unclear. It was initially thought to be the 70kD component of a nucleosome remodeling deacetylase complex, NuRD, but it is more likely that this component is a different but very similar protein. These two proteins are so closely related, though, that they share the same types of domains. These domains include two DNA binding domains, a dimerization domain, and a domain commonly found in proteins that methylate DNA. The profile and activity of this gene product suggest that it is involved in regulating transcription and that this may be accomplished by chromatin remodeling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pericentrin
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone PSH0-66
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene PCNT
Gene Accession No. O95613
Gene Alias AW476095; C86676; KEN; kendrin; KIAA0402; m239Asp; m275Asp; MOPD2; PCN; Pcnt; PCNT2; PCNTB; PCTN2; Pericentrin; pericentrin (kendrin); pericentrin 2; pericentrin-2; pericentrin-250; pericentrin-360; pericentrin-380; pericentrin-B; SCKL4
Gene Symbols PCNT
Host Species Rabbit
Immunogen Recombinant protein within human aa 201-600/3,336.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5116
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.