Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Perifosine
GREENER_CHOICESDP

Catalog No. 608750 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-875-0 50 mg
60-871-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 60-875-0 Supplier R&D Systems™ Supplier No. 6087/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. PKB/Akt inhibitor; also modulates Kv2.1 current

Perifosine is a PKB/Akt inhibitor; accumulates in and reorganizes lipid rafts disrupting Akt signaling. Also inhibits MAPK pathway and induces autophagy. Induces apoptosis in leukemia cell lines. Decreases Kv2.1 current amplitude by modulating its inactivation gating. Antitumor. Orally bioavailable.

Specifications

Chemical Name or Material Perifosine
CAS 157716-52-4
Synonym Octadecyl-(1,1-dimethyl-4-piperidylio) phosphate
Quantity 50 mg
Solubility Soluble to 100mM in water and to 100mM in ethanol
Product Line Tocris
Molecular Formula C25H52NO4P
Formula Weight 461.66
Physical Form Solid
Color White
Product Type Bioactive Small Molecule
Purity >98%
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.