Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perilipin-1 Mouse anti-Human,Bovine, KHL, Clone: Peri 112.17.1, MilliporeSigma™

Catalog No. MABS19422MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS19422MI 25 μg
MABS19421MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABS19422MI Supplier MilliporeSigma Supplier No. MABS194225UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Perilipin-1 Monoclonal antibody specifically detects Perilipin-1 in Human,Bovine samples. It is validated for Electron Microscopy, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Perilipin-1
Applications Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone Peri 112.17.1
Conjugate KLH
Gene Accession No. NP_002657
Gene Alias Lipid droplet-associated protein
Gene Symbols PLIN1, PERI, PLIN
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to first 20 amino acids from the N-terminal region of human Perilipin-1.
Molecular Weight of Antigen ∼65 KDa observed; 55.99 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G Purified
Quantity 25 μg
Research Discipline Signaling
Primary or Secondary Primary
Test Specificity Clone Peri 112.17.1 is a mouse monoclonal antibody that detects Perilipin-1. It targets an epitope within 20 amino acids from the N-terminal region.
Target Species Human, Bovine
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.