Learn More
Perilipin-2/ADFP Antibody (2C5A3), Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP223486SS
 View more versions of this product
Catalog No. NBP223486SS
Only null left
Add to Cart
Description
Description
Perilipin-2/ADFP Monoclonal specifically detects Perilipin-2/ADFP in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
Specifications
Perilipin-2/ADFP
Monoclonal
1.0 mg/mL
Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:10 - 1:100, Immunohistochemistry-Paraffin 1:200 - 1:1000
ADFP, Adipophilin, Adipose differentiation-related proteinadipophilin, ADRP, MGC10598, perilipin 2, perilipin-2
Mouse
Ammonium sulfate precipitation
RUO
Primary
Human
Precipitation
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
2C5A3
Unconjugated
Q99541
PLIN2
Partial recombinant human ADFP (between residues 200-437) expressed in E. coli [UniProt Q99541]
0.025 mL
Cancer, Hypoxia, Lipid and Metabolism, Lipid Droplets, Signal Transduction
123
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only