Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Perilipin-2/ADFP Antibody (ADFP/1366), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP254313AF488
Description
Perilipin-2/ADFP Monoclonal specifically detects Perilipin-2/ADFP in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Perilipin-2/ADFP | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PLIN2 | |
Recombinant fragment (around aa 249-376) of human Perilipin-2/ADFP protein (exact sequence is proprietary) (Uniprot: Q99541) | |
0.1 mL | |
Cancer, Hypoxia, Lipid and Metabolism, Lipid Droplets, Signal Transduction | |
Human | |
IgG2b λ |
ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
ADFP/1366 | |
ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
ADFP, Adipophilin, Adipose differentiation-related proteinadipophilin, ADRP, MGC10598, perilipin 2, perilipin-2 | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction