Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Perilipin-2/ADFP Antibody (ADFP/2755R), DyLight 594, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP308454DL594
Description
Perilipin-2/ADFP Monoclonal antibody specifically detects Perilipin-2/ADFP in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry (Paraffin)Specifications
Perilipin-2/ADFP | |
Monoclonal | |
DyLight 594 | |
ADFP, Adipophilin, Adipose differentiation-related proteinadipophilin, ADRP, MGC10598, perilipin 2, perilipin-2 | |
Recombinant fragment (around aa 249-376) of human Perilipin-2/ADFP protein (exact sequence is proprietary) (Uniprot: Q99541) | |
100 μg | |
Cancer, Cardiovascular Biology, Diabetes Research, Epigenetics, Hypoxia, Lipid and Metabolism, Lipid Droplets, metabolism, Signal Transduction | |
123 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA, Immunohistochemistry (Paraffin) | |
ADFP/2755R | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction