Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Perilipin-2/ADFP Antibody, Alexa Fluor™ 594

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11040878AF594
Description
Perilipin-2/ADFP Polyclonal specifically detects Perilipin-2/ADFP in Human, Mouse, Porcine, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Perilipin-2/ADFP | |
Polyclonal | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin | |
ADFP, Adipophilin, Adipose differentiation-related proteinadipophilin, ADRP, MGC10598, perilipin 2, perilipin-2 | |
Rabbit | |
Affinity Purified | |
Cancer, Hypoxia, Lipid and Metabolism, Lipid Droplets, Signal Transduction | |
123.0 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
PLIN2 | |
A synthetic peptide made to an internal portion of human ADFP (within residues 150-250) [Swiss-ProtNo. Q99541] | |
0.1 mL | |
Primary | |
Human, Mouse, Pig, Monkey | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction