Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Perilipin-2/ADFP Antibody, Biotin

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11040878B
Description
Perilipin-2/ADFP Polyclonal specifically detects Perilipin-2/ADFP in Human, Mouse, Porcine, Monkey samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Perilipin-2/ADFP | |
| Polyclonal | |
| Western Blot, Simple Western, Flow Cytometry 2.5 μg/mL, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| Q99541 | |
| PLIN2 | |
| A synthetic peptide made to an internal portion of human ADFP (within residues 150-250) [Swiss-Prot# Q99541] | |
| 0.1 mL | |
| Cancer, Lipid and Metabolism, Lipid Droplets, Signal Transduction | |
| 123 | |
| Human, Mouse, Pig, Monkey | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| ADFP, Adipophilin, Adipose differentiation-related proteinadipophilin, ADRP, MGC10598, perilipin 2, perilipin-2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human and mouse. Immunogen has 100% identity to monkey, porcine and bovine ADFP, 94% identity tp sheep and 84% identity to rat. | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction