Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perilipin-2/ADFP Antibody - BSA Free, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$247.53 - $603.77

Specifications

Antigen Perilipin-2/ADFP
Concentration 1.17 mg/mL
Dilution Western Blot 2 ug/ml, Simple Western 1:50, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:200, Immunohistochemistry-Frozen reported in scientific literature (PMID 35041644), Dual RNAscope ISH-IHC
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), In Situ Hybridization (ISH)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB11040877T
SDP
View Documents
Novus Biologicals
NB11040877SS
0.025 mL
Each for $247.53
Only null left
Add to Cart
 
NB11040877
SDP
View Documents
Novus Biologicals
NB11040877
0.1 mL
Each for $603.77
Only null left
Add to Cart
 
Description

Description

Perilipin-2/ADFP Polyclonal specifically detects Perilipin-2/ADFP in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Dual RNAscope ISH-IHC.
Specifications

Specifications

Perilipin-2/ADFP
Western Blot 2 ug/ml, Simple Western 1:50, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:200, Immunohistochemistry-Frozen reported in scientific literature (PMID 35041644), Dual RNAscope ISH-IHC
Polyclonal
Rabbit
Cancer, Hypoxia, Lipid and Metabolism, Lipid Droplets, Signal Transduction
PBS and 30% Glycerol with 0.05% Sodium Azide
ADFP, Adipophilin, Adipose differentiation-related proteinadipophilin, ADRP, MGC10598, perilipin 2, perilipin-2
PLIN2
IgG
Affinity Purified
Human and mouse. Immunogen has 88% sequence identity with rat ADFP. Customer feedback has been negative on trout liver.
1.17 mg/mL
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), In Situ Hybridization (ISH)
Unconjugated
RUO
Human, Mouse, Rat, Bovine
Q99541, P43883
123
A synthetic peptide made to a C-terminal region of mouse ADFP (within residues 350-425) [Swiss-Prot# P43883]
Primary
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
51 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.