Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perilipin-2/ADFP Antibody - BSA Free
SDP

Rabbit Polyclonal Antibody has been used in 3 publications

$220.00

Specifications

Antigen Perilipin-2/ADFP
Concentration 1.23 mg/ml
Dilution Western Blot 2 μg/mL, Simple Western 1:50, Flow Cytometry 2.5 μg/mL, Immunohistochemistry 1:200, Immunohistochemistry-Paraffin 1:200, Immunohistochemistry-Frozen
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB11040878T
SDP
View Documents
Novus Biologicals
NB11040878SS
0.025 mL
Each for $220.00
Only null left
Add to Cart
 
NB11040878
SDP
View Documents
Novus Biologicals
NB11040878
0.1 mL N/A N/A N/A
Description

Description

Perilipin-2/ADFP Polyclonal specifically detects Perilipin-2/ADFP in Human, Mouse, Porcine, Monkey samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications

Specifications

Perilipin-2/ADFP
Western Blot 2 μg/mL, Simple Western 1:50, Flow Cytometry 2.5 μg/mL, Immunohistochemistry 1:200, Immunohistochemistry-Paraffin 1:200, Immunohistochemistry-Frozen
Polyclonal
Rabbit
Cancer, Lipid and Metabolism, Lipid Droplets, Signal Transduction
PBS, 30% Glycerol with 0.05% Sodium Azide
ADFP, Adipophilin, Adipose differentiation-related proteinadipophilin, ADRP, MGC10598, perilipin 2, perilipin-2
PLIN2
IgG
Affinity Purified
Human and mouse. Immunogen has 100% identity to monkey, porcine and bovine ADFP, 94% identity tp sheep and 84% identity to rat.
1.23 mg/ml
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Unconjugated
RUO
Human, Mouse, Pig, Monkey
Q99541
123
A synthetic peptide made to an internal portion of human ADFP (within residues 150-250) [Swiss-Prot# Q99541]
Primary
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.