Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Perilipin-2/ADFP Antibody (ADFP/1493) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$646.00
Specifications
| Antigen | Perilipin-2/ADFP |
|---|---|
| Clone | ADFP/1493 |
| Concentration | 1 mg/mL |
| Dilution | ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml |
| Applications | ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB254312100
![]() |
Novus Biologicals
NBP254312100UG |
100 μg |
Each for $646.00
|
|
|||||
NB254312200
![]() |
Novus Biologicals
NBP254312200UG |
200 μg | N/A | N/A | N/A | ||||
Description
Perilipin-2/ADFP Monoclonal specifically detects Perilipin-2/ADFP in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Perilipin-2/ADFP | |
| 1 mg/mL | |
| ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer, Hypoxia, Lipid and Metabolism, Lipid Droplets, Signal Transduction | |
| PBS with No Preservative | |
| 123 | |
| Recombinant human Adipophilin protein fragment (aa249-376) (exact sequence is proprietary) | |
| Primary | |
| Store at -20 to -80C. Avoid freeze-thaw cycles. |
| ADFP/1493 | |
| ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| ADFP, Adipophilin, Adipose differentiation-related proteinadipophilin, ADRP, MGC10598, perilipin 2, perilipin-2 | |
| PLIN2 | |
| IgG2b λ | |
| Protein A or G purified | |
| 48 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title