Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Perilipin-3/TIP47 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11040764MFV610
Description
Perilipin-3/TIP47 Polyclonal antibody specifically detects Perilipin-3/TIP47 in Human, Mouse, Porcine, Primate samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
Perilipin-3/TIP47 | |
Polyclonal | |
Western Blot, Immunocytochemistry/ Immunofluorescence | |
Cargo selection protein TIP47, M6PRBP1MGC2012, Mannose-6-phosphate receptor-binding protein 1, perilipin 3,47 kDa MPR-binding protein, perilipin-3, Placental protein 17,47 kDa mannose 6-phosphate receptor-binding protein, PP17MGC11117, tail-interacting protein, 47 kD, TIP47mannose-6-phosphate receptor binding protein 1 | |
A synthetic peptide made to a region within the C-terminus (within residues 350-435) of the human TIP47 protein. [Swiss-Prot# O60664] | |
0.1 mL | |
Diabetes Research, Golgi Apparatus Markers, Lipid and Metabolism, Lipid Droplets, Signal Transduction | |
10226 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunocytochemistry | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Pig, Primate | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction