Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perilipin-3/TIP47 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.50 - $499.50

Specifications

Antigen Perilipin-3/TIP47
Concentration 1.00 mg/mL
Dilution Western Blot 2 ug/ml, Immunocytochemistry/Immunofluorescence 1:100
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB11040764T
SDP
View Documents
Novus Biologicals
NB11040764SS
0.025 mL
Each for $206.50
Only null left
Add to Cart
 
NB11040764
SDP
View Documents
Novus Biologicals
NB11040764
0.1 mL
Each for $499.50
Only null left
Add to Cart
 
Description

Description

Perilipin-3/TIP47 Polyclonal specifically detects Perilipin-3/TIP47 in Human, Mouse, Porcine, Primate samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Perilipin-3/TIP47
Western Blot 2 ug/ml, Immunocytochemistry/Immunofluorescence 1:100
Polyclonal
Rabbit
Diabetes Research, Golgi Apparatus Markers, Lipid and Metabolism, Lipid Droplets, Signal Transduction
Tris-Glycine and 0.15M NaCl with 0.05% Sodium Azide
Cargo selection protein TIP47, M6PRBP1MGC2012, Mannose-6-phosphate receptor-binding protein 1, perilipin 3,47 kDa MPR-binding protein, perilipin-3, Placental protein 17,47 kDa mannose 6-phosphate receptor-binding protein, PP17MGC11117, tail-interacting protein, 47 kD, TIP47mannose-6-phosphate receptor binding protein 1
PLIN3
IgG
Affinity Purified
1.00 mg/mL
Western Blot, Immunocytochemistry, Immunofluorescence
Unconjugated
RUO
Human, Mouse, Pig, Primate
O60664
10226
A synthetic peptide made to a region within the C-terminus (within residues 350-435) of the human TIP47 protein. [Swiss-Prot# O60664]
Primary
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.