Learn More
Invitrogen™ Perilipin A/B Polyclonal Antibody

Description
PA1-1052 detects perilipin A and B from rat and 3T3-L1 cell extract. PA1-1052 has been successfully used in Western blot procedures. By Western blot, this antibody detects ~62 and 46 kDa proteins representing perilipin A and B, respectively from 3T3-L1 cell extract The PA1-1052 immunogen is a synthetic peptide corresponding to residues M(1) S M N K G P T L L D G D L P E Q(17) C of rat Perilipin A and B. This sequence is 82% conserved in humans. The PA1-1052 immunizing peptide (Cat. # PEP-170) is available for use in neutralization and control experiments.
Specifications
Specifications
| Antigen | Perilipin A/B |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% sodium azide |
| Gene | Plin2 |
| Gene Accession No. | P43883, P43884, Q8CGN5 |
| Gene Alias | 6030432J05Rik; AA407157; ADFP; Adipophilin; adipose differentiation related protein; adipose differentiation-related protein; ADPH; ADRP; FPLD4; lipid droplet-associated protein; PERI; PERIA; perilipin 1; perilipin 2; perilipin A; perilipin B; perilipin-1; Perilipin-2; PLIN; PLIN1; Plin2 |
| Gene Symbols | Plin1, Plin2 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.