Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Perilipin Antibody, DyLight 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299070DL594
Description
Perilipin Polyclonal specifically detects Perilipin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Perilipin | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
PERI, perilipin, perilipin 1, perilipin-1, PLINLipid droplet-associated protein | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human Perilipin. | |
0.1 mL | |
Diabetes Research, Lipid and Metabolism, Lipid Droplets | |
5346 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 594 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction