Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Periodic acid, 99+%, extra pure

Catalog No. AC198400025
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg

Periodic acid, 99+%, H5IO6, CAS Number-10450-60-9, pentahydroxidooxidoiodine, paraperiodic acid, orthoperiodic acid, acid, periodic, hexaoxoiodic 5-acid, tetrahydroxyiodous acid, pentahydroxy-l5-iodane oxide, periodic acid, periodic acid, cryst, unii-ak1d44l87g, 100g, 1.0% max. Iodic Acid | CAS: 10450-60-9 | H5IO6 | 227.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10450-60-9
Molecular Formula H5IO6
Molecular Weight (g/mol) 227.94
MDL Number MFCD00011344
InChI Key TWLXDPFBEPBAQB-UHFFFAOYSA-N
Synonym orthoperiodic acid, periodic acid, tetrahydroxyiodous acid, acid, periodic, paraperiodic acid, pentahydroxy-l5-iodane oxide, unii-ak1d44l87g, pentahydroxidooxidoiodine, hexaoxoiodic 5-acid, periodic acid, cryst
PubChem CID 25289
ChEBI CHEBI:29150
SMILES O[I](O)(O)(O)(O)=O

Specifications

Color White to Yellow
Melting Point 122.0°C
Identification Positive for periodate
Packaging Glass bottle
Assay Percent Range 99% min. (Iodometry)
Quantity 2.5 kg
Physical Form Crystals or Crystalline Powder
Linear Formula H5IO6
Fieser 01,815; 02,313; 03,220; 04,374; 05,508; 10,304; 13,238; 16,272
Merck Index 15, 7286
Solubility Information Solubility in water: 3 kg/l (20°C). Other solubilities: soluble in alcohol,slightly soluble in ether
IUPAC Name pentahydroxy(oxo)-$l^{7}-iodane
Formula Weight 227.94
Percent Purity ≥99%
Grade Extra Pure
Chemical Name or Material Periodic acid
Show More Show Less
Hazard Statement GHS H Statement
May cause fire or explosion; strong oxidizer.
Causes severe skin burns and eye damage.
Causes damage to organs through prolonged or repeated exposure.
Precautionary Statement GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 233-937-

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.