Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Periodic acid, 99+%, extra pure, Acros Organics

Click to view available options
Quantity:
5g
100g
500g
2.5kg
10450-60-9
H5IO6
227.94
TWLXDPFBEPBAQB-UHFFFAOYSA-N
orthoperiodic acid,periodic acid,tetrahydroxyiodous acid,acid, periodic,paraperiodic acid,pentahydroxy-l5-iodane oxide,unii-ak1d44l87g,pentahydroxidooxidoiodine,hexaoxoiodic 5-acid,periodic acid, cryst
25289
CHEBI:29150
pentahydroxy(oxo)-$l^{7}-iodane
OI(=O)(O)(O)(O)O
10450-60-9
H5IO6
227.94
TWLXDPFBEPBAQB-UHFFFAOYSA-N
orthoperiodic acid,periodic acid,tetrahydroxyiodous acid,acid, periodic,paraperiodic acid,pentahydroxy-l5-iodane oxide,unii-ak1d44l87g,pentahydroxidooxidoiodine,hexaoxoiodic 5-acid,periodic acid, cryst
25289
CHEBI:29150
pentahydroxy(oxo)-$l^{7}-iodane
OI(=O)(O)(O)(O)O
Additional Information 1.0% max. Iodic Acid
Assay Percent Range 99% min. (Iodometry)
Linear Formula H5IO6
Fieser 01,815; 02,313; 03,220; 04,374; 05,508; 10,304; 13,238; 16,272
Merck Index 15, 7286
Solubility Information Solubility in water: 3 kg/l (20°C). Other solubilities: soluble in alcohol,slightly soluble in ether
Formula Weight 227.94
Physical Form Crystals or Crystalline Powder
Percent Purity 99+%
Grade Extra Pure
Bromide 200ppm max. (Bromide, Bromate (as Cl))
Chloride and Chlorate 200ppm max. (as Cl)
Identification Positive for periodate
Insoluble in Water 0.01% max.
Insoluble Matter 0.01% max. (in water)
Iodide 10ppm max.
Iron (Fe) 50ppm max.
Nitrate 100ppm max.
Packaging Glass bottle
Solubility Solubility in water: 3 kg/l (20°C). Other solubilities: soluble in alcohol, slightly soluble in ether
Sulfate 200ppm max.
Sulfated Ash 0.05% max.
Color White to Yellow
Melting Point 122.0°C
Quantity 500g
Chemical Name or Material Periodic acid
Show More Show Less

GHS H Statement
May cause fire or explosion; strong oxidizer.
Causes severe skin burns and eye damage.
Causes damage to organs through prolonged or repeated exposure.

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 233-937-