Learn More
Periodic Acid Solution, in Acetic Acid, Reagents
Specifications
Specifications
| Chemical Name or Material | Periodic Acid Solution, in Acetic Acid |
| pH | <2 |
| Color | Colorless to Light Yellow |
| CAS | 64-19-7, 7732-18-5, 10450-60-9 |
| Boiling Point | 117°C |
| Packaging | Glass Bottle |
| Specific Gravity | 1.0477 |
| Quantity | 1 L |
| Physical Form | Liquid |
| Synonym | Iodine Tetraoxide |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.