Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Periodic Acid (White to Pale-Yellow Crystals or White Powder), Fisher BioReagents

Catalog No. p-27552 Shop All Fisherbrand Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Poly Bottle
Periodic Acid is used to decompose organic compounds.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10450-60-9
Molecular Formula H5IO6
Molecular Weight (g/mol) 227.94
MDL Number MFCD00011344
InChI Key TWLXDPFBEPBAQB-UHFFFAOYSA-N
Synonym orthoperiodic acid, periodic acid, tetrahydroxyiodous acid, acid, periodic, paraperiodic acid, pentahydroxy-l5-iodane oxide, unii-ak1d44l87g, pentahydroxidooxidoiodine, hexaoxoiodic 5-acid, periodic acid, cryst
PubChem CID 25289
ChEBI CHEBI:29150
SMILES O[I](O)(O)(O)(O)=O

Specifications

pH 1
Color White
Melting Point 122°C
Density 1.4 g/cm3
Heavy Metals (as Pb) 0.005% max.
Identification Pass Test
Packaging Poly Bottle
Assay Percent Range ≥99 %
Quantity 25 g
Physical Form Solid
Linear Formula HIO4·2H2O
Merck Index 15, 7286
IUPAC Name tetrahydroxyiodous acid
Formula Weight 227.94
Percent Purity ≥99%
Chemical Name or Material Periodic Acid
Show More Show Less
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Oxidizer: Contact with combustible/organic material may cause fire. Use personal protective equipment. Keep away from clothing and other combustible materials. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting.

NFPA

Health:3
Flammability:0
Instability:2
DOTInformation DOT Class 5.1, : Oxidizer
Recommended Storage RT
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.