Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Periostin Monoclonal Antibody (C2), Invitrogen™

Catalog No. FEMA550892
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550892 20 μL
FEMA550891 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FEMA550892 Supplier Invitrogen™ Supplier No. MA550892
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Periostin (PN), also designated osteoblast-specific factor 2 (OSF-2), is a disulfide linked protein originally isolated as a osteoblast-specific factor. Periostin is a secreted protein that binds heparin and functions as a ligand for alpha(V)beta(3) and alpha(V)beta(5) integrins. In preosteoblasts, Periostin acts as a cell adhesion molecule and plays a role in osteoblast recruitment, spreading and attachment. Periostin is mainly detected in lower gastrointestinal tract, aorta, stomach, placenta, uterus and breast tissues but is up-regulated in epithelial ovarian tumors and overexpressed in breast cancer. Expression of Periostin is increased by bone morphogenetic protein (BMP2) and transforming growth factor beta 1(TGF beta 1). Periostin contains a typical signal sequence, followed by a cysteine-rich domain, a fourfold repeated domain, which shows homology with the insect protein fasciclin, and a C-terminal domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Periostin
Applications Western Blot, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone C2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene Postn
Gene Accession No. Q15063
Gene Alias A630052E07Rik; AI747096; fasciclin I-like; Fasciclin-I like; LOW QUALITY PROTEIN: periostin; MGC119510; MGC119511; OSF 2; Osf2; OSF-2; osteoblast specific factor 2; osteoblast specific factor 2 (fasciclin I-like); osteoblast-specific factor 2; PDLPOSTN; peri; periodontal ligament-specific periostin; periostin; periostin variant 1; periostin variant 2; periostin variant 3; periostin variant 4; periostin variant 5; periostin variant 6; periostin variant 7; periostin variant 8; periostin variant 9; periostin, osteoblast specific factor; periostin-like factor; Plf; PN; POSTN; POSTN protein; RP11 412K4.1; RP11-412K4.1
Gene Symbols Postn
Host Species Mouse
Immunogen Recombinant Human POSTN protein, Pro97-Leu230 (Accession #Q15063).
Purification Method Protein A/G
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10631, 361945
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.