Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Periostin Monoclonal Antibody (OTI2D10), TrueMAB™, OriGene
SDP

Catalog No. 501679896
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-9896 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-9896 Supplier OriGene Supplier No. TA803713
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Enhances incorporation of BMP1 in the fibronectin matrix of connective tissues, and subsequent proteolytic activation of lysyl oxidase LOX. Induces cell attachment and spreading and plays a role in cell adhesion. May play a role in extracellular matrix mineralization.

Specifications

Antigen Periostin
Applications Western Blot
Classification Monoclonal
Clone OTI2D10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene POSTN
Gene Accession No. Q15063
Gene Alias OSF-2, OSF2, PDLPOSTN, PN
Gene Symbols POSTN
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 538-781 of human POSTN produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10631
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.