Learn More
Periostin Mouse anti-Human, Clone: OTI2F7, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF803712

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Enhances incorporation of BMP1 in the fibronectin matrix of connective tissues, and subsequent proteolytic activation of lysyl oxidase LOX. Induces cell attachment and spreading and plays a role in cell adhesion. May play a role in extracellular matrix mineralization.Specifications
Periostin | |
Monoclonal | |
Unconjugated | |
POSTN | |
OSF-2, OSF2, PDLPOSTN, PN | |
Mouse | |
Affinity Chromatography | |
RUO | |
10631 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI2F7 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q15063 | |
POSTN | |
Human recombit protein fragment corresponding to amino acids 538-781 of human POSTN produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.