Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Periostin, Mouse, Unlabeled, Clone: 5B2.3,

Catalog No. MABS1183MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABS1183MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS1183MI Supplier MilliporeSigma™ Supplier No. MABS1183
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Periostin (UniProt Q15063; also known as OSF-2, Osteoblast-specific factor 2, PN) is encoded by the POSTN (also known as PSF2) gene (Gene ID 10631) in human. Periostin is an extracellular matrix (ECM) protein predominantly expressed in collagen-rich fibrous connective tissues, including heart valves, tendons, perichondrium, periosteum, and periodontal ligament (PDL). Periostin plays an essential role in the process of remodeling during tissue/organ development/repair and inflammation. Periostin derived from fibroblasts is shown to induce keratinocytes differentiation and proliferation. Periostin is a mediator for epidermal hyperplasia and is highly expressed in the dermis of both atopic dermatitis (AD) and psoriasis patients. Periostin level is also reported to be upregulated in patients with acute or chronic hepatitis and correlates well with serum TGF- 1 and TGF- 2 levels. The PDL-POSTN spliced isoform is predominantly expressed in PDL, it binds integrin v 3 with a higher affinity and induces strong activation of the integrin v 3-focal adhesion kinase (FAK) signaling pathway. Periostin is produced with a signal peptide (a.a. 1-21), the removal of which yields the mature protein with an N-terminal cysteine-rich EMI (a.a. 40-94) domain and four asciclin I or FAS1 domains (a.a. 97-230, 234-365, 368-492, 496-628). Alternative splicing generate several C-terminal domain variants with tissue-specific expression patterns.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Periostin
Applications Western Blot
Classification Monoclonal
Clone 5B2.3
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG2a in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene POSTN, OSF2
Gene Accession No. Q15063
Host Species Mouse
Immunogen GST-tagged recombinant human periostin C-terminal fragment.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_006466
Test Specificity Clone 5B2.3 epitope lies within a region C-terminal to the FAS1 domains. Immunogen covered region is 100% present in isoforms 1 (OSF-2OS), 2 (OSF-2p1), and 5, but partially missing in spliced isoforms 3, 4 (PDL-POSTN), 6, and 7. Clone 5B2.3 immunoreactivity toward these latter isoforms is possible, but has not been confirmed.
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.