Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Periostin/OSF-2 Antibody (031), DyLight 650

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290594C
Description
Periostin/OSF-2 Monoclonal specifically detects Periostin/OSF-2 in Mouse samples. It is validated for ELISA.Specifications
Periostin/OSF-2 | |
Monoclonal | |
DyLight 650 | |
MGC119510, MGC119511, OSF-2osteoblast specific factor 2 (fasciclin I-like), OSF2periodontal ligament-specific periostin, Osteoblast-specific factor 2, PDLPOSTN, periostin, periostin isoform thy2, periostin isoform thy4, periostin isoform thy6, periostin isoform thy8, periostin, osteoblast specific factor, PNRP11-412K4.1 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Periostin/OSF-2 (Met 1-Gln 811; NP_056599.1) . | |
0.1 mL | |
Cardiovascular Biology, Extracellular Matrix | |
10631 | |
Mouse | |
Purified |
ELISA | |
31 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
No cross-reactivity in ELISA with: Human POSTN | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction