Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Periostin/OSF-2 Antibody (OTI2B2), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273336JF646
Description
Periostin/OSF-2 Monoclonal specifically detects Periostin/OSF-2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Periostin/OSF-2 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
POSTN | |
Recombinant protein fragment corresponding to amino acids 538-781 of human Periostin/OSF-2 (NP_001129407) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Western Blot, Immunohistochemistry | |
OTI2B2 | |
Western Blot, Immunohistochemistry | |
MGC119510, MGC119511, OSF-2osteoblast specific factor 2 (fasciclin I-like), OSF2periodontal ligament-specific periostin, Osteoblast-specific factor 2, PDLPOSTN, periostin, periostin isoform thy2, periostin isoform thy4, periostin isoform thy6, periostin isoform thy8, periostin, osteoblast specific factor, PNRP11-412K4.1 | |
Mouse | |
Affinity Purified | |
Cardiovascular Biology, Extracellular Matrix | |
10631.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction