Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Periostin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582458
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582458 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582458 Supplier Invitrogen™ Supplier No. PA582458
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GLFINHYPNG VVTVNCARII HGNQIATNGV VHVIDRVLTQ IGTSIQDFIE AEDDLSSFRA AAITSDILEA LGRDGHFTLF APTNEAFEKL PRGVLERIMG DKVASEALMK YHILNTLQCS ESIMGGAVFE TLEG Highest antigen sequence indentity to the following orthologs: Rat - 95%, Mouse - 95%.

Periostin (PN), also designated osteoblast-specific factor 2 (OSF-2), is a disulfide linked protein originally isolated as a osteoblast-specific factor. Periostin is a secreted protein that binds heparin and functions as a ligand for alpha(V)beta(3) and alpha(V)beta(5) integrins. In preosteoblasts, Periostin acts as a cell adhesion molecule and plays a role in osteoblast recruitment, spreading and attachment. Periostin is mainly detected in lower gastrointestinal tract, aorta, stomach, placenta, uterus and breast tissues but is up-regulated in epithelial ovarian tumors and overexpressed in breast cancer. Expression of Periostin is increased by bone morphogenetic protein (BMP2) and transforming growth factor beta 1(TGF beta 1). Periostin contains a typical signal sequence, followed by a cysteine-rich domain, a fourfold repeated domain, which shows homology with the insect protein fasciclin, and a C-terminal domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Periostin
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Postn
Gene Accession No. Q15063
Gene Alias A630052E07Rik; AI747096; fasciclin I-like; Fasciclin-I like; LOW QUALITY PROTEIN: periostin; MGC119510; MGC119511; OSF 2; Osf2; OSF-2; osteoblast specific factor 2; osteoblast specific factor 2 (fasciclin I-like); osteoblast-specific factor 2; PDLPOSTN; peri; periodontal ligament-specific periostin; periostin; periostin variant 1; periostin variant 2; periostin variant 3; periostin variant 4; periostin variant 5; periostin variant 6; periostin variant 7; periostin variant 8; periostin variant 9; periostin, osteoblast specific factor; periostin-like factor; Plf; PN; POSTN; POSTN protein; RP11 412K4.1; RP11-412K4.1
Gene Symbols Postn
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human POSTN. Recombinant protein control fragment (Product #RP-89721).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10631
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.