Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Peripheral Node Addressin Antibody (MECA-79) - Low Endotoxin and Azide Free, Novus Biologicals™
SDP

Catalog No. p-7163489 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB001660 0.1 mg
NB001659 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB001660 Supplier Novus Biologicals Supplier No. NBP278790SS
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Peripheral Node Addressin Monoclonal specifically detects Peripheral Node Addressin in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, In vivo assay.

Specifications

Antigen Peripheral Node Addressin
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone MECA-79
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:100-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:100-1:500, Immunohistochemistry-Frozen 1:100-1:500, In vivo assay reported in scientific literature (PMID 30277476)
Formulation PBS with No Preservative
Gene Alias MECA-79, Peripheral Node Addressin, PNAd
Host Species Rat
Immunogen Mouse lymph node stromal cells
Purification Method IgM purified
Quantity 0.1 mg
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.