Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Peripherin Antibody (8G2), Novus Biologicals™
SDP

Catalog No. NB018135 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018135 0.025 mL
N300138100U 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018135 Supplier Novus Biologicals Supplier No. NB3001380.025ML
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peripherin Monoclonal specifically detects Peripherin in Human, Mouse, Rat, Porcine, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Peripherin
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Classification Monoclonal
Clone 8G2
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:5000, Immunohistochemistry 1:500, Immunocytochemistry/Immunofluorescence 1:500
Formulation PBS, 50% Glycerol with 0.05% sodium azide
Gene Alias Neurofilament 4, neurofilament 4 (57kD), peripherin, PRPH1NEF4
Gene Symbols PRPH
Host Species Mouse
Immunogen Recombinant rat Peripherin purified from E. coli. [UniProtNo. P21807]
Quantity 0.025 mL
Primary or Secondary Primary
Gene ID (Entrez) 5630
Target Species Rat, Bovine, Mouse, Pig, Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.