Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Peripherin Antibody (OTI1E10), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273337JF549
Description
Peripherin Monoclonal specifically detects Peripherin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Peripherin | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PRPH | |
Full length recombinant protein of human Peripherin (NP_006253) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI1E10 | |
Western Blot, Immunohistochemistry | |
Neurofilament 4, neurofilament 4 (57kD), peripherin, PRPH1NEF4 | |
Mouse | |
Affinity Purified | |
Cancer, Cytoskeleton Markers, Neuronal Cell Markers, Neuroscience, Signal Transduction | |
5630.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction