Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Peripherin Mouse anti-Rat, Mouse, Human, Clone: 7C5, MilliporeSigma™

Catalog No. MAB5380I100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5380I100 100 μg
MAB5380I25 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MAB5380I100 Supplier MilliporeSigma Supplier No. MAB5380I100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peripherin Monoclonal antibody specifically detects Peripherin in Rat, Mouse, Human samples. It is validated for Immunocytochemistry, Immunofluorescence, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Peripherin
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 7C5
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG1 in PBS with 5 mM sodium azide with 50% glycerol.
Host Species Mouse
Immunogen Full length recombinant Rat peripherin expressed in and purified from E. coli.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Rat, Mouse, Human
Content And Storage Stable for 1 year at -10°C to -25°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.