Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Peripherin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143159
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143159 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5143159 Supplier Invitrogen™ Supplier No. PA5143159
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Additional Information: This antibody is expected to recognize all reported isoforms (NP_038667.2; NP_001157060.1; NP_001157061.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.

Peripherin is a 57kD type III intermediate filament that is a specific marker for peripheral neurons, including enteric ganglion cells. Peripherin is expressed in the developing peripheral nervous system and is highly enriched in neuronal derivatives of the neural crest. Peripherin offers an advantage over other neural markers, such as S100 and NSE in that it does not stain chrmaffin cell types.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Peripherin
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene PRPH
Gene Accession No. P15331, P21807
Gene Alias if3; nef4; Neurofilament 4; neurofilament 4 (57kD); neuronal intermediate filament IF3; Perf; Peripherin; peripherin 1; peripherin L homeolog; plasticin; PRPH; prph.L; PRPH1; XELAEV_18013332mg; XIF3
Gene Symbols PRPH
Host Species Goat
Immunogen Peptide with sequence C-QKEQHSDLDKSSIH.
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19132, 24688
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.