Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PERK Chicken anti-Human, Mouse, Rat, Polyclonal, FabGennix
Chicken Polyclonal Antibody
Supplier: Fabgennix Inc PERK101Y

Description
PERK Polyclonal antibody specifically detects PERK in Human, Mouse, Rat samples. It is validated for Dot Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Western BlotSpecifications
PERK | |
Polyclonal | |
Unconjugated | |
EIF2AK3 | |
2.7.11.1; AI427929; EIF2AK3; eukaryotic translation initiation factor 2 alpha kinase 3; eukaryotic translation initiation factor 2-alpha kinase 3; HsPEK; OTTHUMP00000207187; OTTHUMP00000207188; Pancreatic eIF2-alpha kinase; PEK; Perk; PRKR-like endoplasmic reticulum kinase; WRS | |
Synthetic peptide taken within amino acid region 1000-1050 on human Eukaryotic translation initiation factor 2-alpha kinase 3 protein. | |
200 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgY |
Dot Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
0.5-1.5 mg/mL | |
proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8 | |
Q9NZJ5, Q9Z1Z1, Q9Z2B5 | |
Chicken | |
Affinity chromatography | |
RUO | |
13666, 29702, 9451 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction