Learn More
Invitrogen™ PERK Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA579193
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, human COLO-320 whole cell, human A549 whole cell, human SK-OV-3 whole cell, Human A431 whole cell, rat brain tissue, mouse brain tissue. Flow: HepG2 cell.
EIF2AK3 (PERK) is one of 4 kinases that specifically phosphorylate Ser51 of translation initiation factor eIF2-alpha in response to various environmental stresses, leading to a decrease in protein sythesis. In the case of EIF2AK3, signaling is initiated by misfolded proteins in the ER. Defects in EIF2AK3 are the cause of Wolcott-Rallison syndrome (WRS), also known as multiple epiphyseal dysplasia with early-onset diabetes mellitus. WRS is a rare autosomal recessive disorder, characterized by permanent neonatal or early infancy insulin-dependent diabetes and, at a later age, epiphyseal dysplasia, osteoporosis, growth retardation and other multisystem manifestations, such as hepatic and renal dysfunctions, mental retardation and cardiovascular abnormalities.
Specifications
| PERK | |
| Polyclonal | |
| Unconjugated | |
| EIF2AK3 | |
| 2.7.11.1; AI427929; EIF2AK3; eukaryotic translation initiation factor 2 alpha kinase 3; eukaryotic translation initiation factor 2-alpha kinase 3; HsPEK; OTTHUMP00000207187; OTTHUMP00000207188; Pancreatic eIF2-alpha kinase; PEK; Perk; PRKR-like endoplasmic reticulum kinase; WRS | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 13666, 29702, 9451 | |
| -20°C | |
| Lyophilized |
| ELISA, Flow Cytometry, Western Blot | |
| 500 μg/mL | |
| PBS with 4mg trehalose and 0.05mg sodium azide | |
| Q9NZJ5, Q9Z1Z1, Q9Z2B5 | |
| EIF2AK3 | |
| E. coli-derived human PERK recombinant protein (Position: R222-Q334). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.