Learn More
Invitrogen™ Perlecan Monoclonal Antibody (A71)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ CSI0017102
Description
CSI 001-71-02 is highly specific for perlecan. There is no evidence for cross-reactivity with other connective tissue proteins (vitronectin, fibronectin, elastin, collagen, laminin). CSI 001-71-02 cross-reacts with human thrombospondin. Other species have not been tested. CSI 001-71-02 has been successfully used in ELISA, immunohistochemistry and Western blotting techniques. CSI 001-71-02 can be used in immunoprecipitation, ELISA and immunostaining of frozen PLP-fixed sections of bovine and human tissues. CSI 001-71-02 can be used in affinity chromatography to separate recombinant domain I from tissue culture fluid. Epitope of CSI 001-71-02 is located in domain I. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL.
Perlecan is a major heparan-sulfate proteoglycan (HSPG) found within all basement membranes and cell surfaces. Because of its strategic location and ability to store and protect growth factors, perlecan has been strongly implicated in the control of tumor cell growth and metastatic behavior. Perlecan possesses angiogenic and growth-promoting attributes primarily by acting as a coreceptor for basic fibroblast growth factor (FGF-2). Suppression of perlecan causes substantial inhibition of neoplastic growth and neovascularization. Thus, perlecan is a potent inducer of neoplasm growth and angiogenesis in vivo and therapeutic interventions targeting this key modulator of tumor progression may improve neoplastic treatment.
Specifications
Perlecan | |
Monoclonal | |
1.03 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
P98160 | |
HSPG2 | |
Bovine corneal endothelial cells. | |
400 μL | |
Primary | |
Human, Bovine | |
Antibody | |
IgG1 κ |
ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
A71 | |
Unconjugated | |
HSPG2 | |
AI852380; Basement membrane-specific heparan sulfate proteoglycan core protein; Endorepellin; endorepellin (domain V region); heparan sulfate proteoglycan 2; HSPG; Hspg2; LG3 peptide; LOC313641; Pcn; per; perlecan; perlecan (heparan sulfate proteoglycan 2); perlecan proteoglycan; PLC; PRCAN; RP11-132G19.2; SJA; SJS; SJS1 | |
Mouse | |
Protein A/G | |
RUO | |
3339, 444872 | |
4°C, store in dark | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.