Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Permanent Orange 90.0+%, TCI America™
SDP

Catalog No. P058725G
Click to view available options
Quantity:
25 g

Chemical Identifiers

CAS 3468-63-1
Molecular Formula C16H10N4O5
Molecular Weight (g/mol) 338.28
MDL Number MFCD00059524
InChI Key JABZBOSSCDRKEA-VLGSPTGOSA-N
Synonym 1-(2,4-Dinitrophenylazo)-2-naphthol, Pigment Orange 5
PubChem CID 9570132
IUPAC Name (1Z)-1-[2-(2,4-dinitrophenyl)hydrazin-1-ylidene]-1,2-dihydronaphthalen-2-one
SMILES [O-][N+](=O)C1=CC=C(N\N=C2/C(=O)C=CC3=CC=CC=C23)C(=C1)[N+]([O-])=O

Specifications

Melting Point 306°C
Quantity 25 g
Formula Weight 338.28
Percent Purity ≥93.0% (E)
Physical Form Crystalline Powder
Chemical Name or Material Permanent Orange
EINECSNumber (5)-3192
RTECSNumber QL3854000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.