Learn More
Peroxidase (Horseradish) Polyclonal Antibody, Biotin, Rockland™

Rabbit Polyclonal Antibody
$178.35 - $3071.52
Specifications
| Antigen | Peroxidase (Horseradish) |
|---|---|
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Biotin |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-1156
![]()
|
Rockland Immunochemicals
20046380100 |
100 μg |
Each for $444.03
|
|
|||||
50-199-1157
![]()
|
Rockland Immunochemicals
2004638S |
25 μL |
Each for $178.35
|
|
|||||
50-199-1155
![]()
|
Rockland Immunochemicals
2004638 |
10 mg |
Each for $3,071.52
|
|
|||||
Description
Store vial at 4°C prior to restoration. For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Anti-Peroxidase (Horseradish) Antibody Biotin Conjugated is an IgG fraction antibody purified from monospecific antiserum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Biotin, anti-Rabbit Serum as well as purified and partially purified Peroxidase [Horseradish]. Cross reactivity against Peroxidase from other sources is unknown.
Chemiluminescent detection systems have emerged as the best all-around method for detection of Western blots. They eliminate the hazards associated with radioactive materials and toxic chromogenic substrates. The speed and sensitivity of these methods are unequalled by traditional alternatives. Because results are generated on film, it is possible to record and store data permanently, and blots detected with chemiluminescent methods are easily stripped for subsequent reprobing with additional antibodies. HRP (Horseradish Peroxidase) conjugated secondary antibodies are utilized in conjunction with specific chemiluminescent substrates to generate the light signal. HRP-antibody conjugates have a very high turnover rate, giving good sensitivity with short reaction times.Specifications
| Peroxidase (Horseradish) | |
| Immunohistochemistry, Immunoprecipitation, Western Blot | |
| Biotin | |
| Rabbit | |
| 0.02M potassium phosphate with 0.15M NaCl, 10 mg/mL BSA and 0.01% sodium azide; pH 7.2 | |
| Peroxidase [Horseradish] | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| 1 mg/mL | |
| Polyclonal | |
| Lyophilized | |
| RUO | |
| Horseradish Peroxidase | |
| Primary | |
| Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.