Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PeroxidaseGoat anti-Tag, Polyclonal, Rockland™
SDP

Catalog No. 501989993
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9993 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9993 Supplier Rockland Immunochemicals Supplier No. 1001138
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Store vial at 4°C prior to restoration. For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-goat serum, purified and partially purified Peroxidase [Horseradish]. Cross reactivity against Peroxidase from other tissues and species may occur but have not been specifically determined.

Chemiluminescent detection systems have emerged as the best all-around method for detection of Western blots. They eliminate the hazards associated with radioactive materials and toxic chromogenic substrates. The speed and sensitivity of these methods are unequalled by traditional alternatives. Because results are generated on film, it is possible to record and store data permanently, and blots detected with chemiluminescent methods are easily stripped for subsequent reprobing with additional antibodies. HRP (Horseradish Peroxidase) conjugated secondary antibodies are utilized in conjunction with specific chemiluminescent substrates to generate the light signal. HRP-antibody conjugates have a very high turnover rate, giving good sensitivity with short reaction times.

Specifications

Antigen Peroxidase
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 45 mg/mL
Conjugate Unconjugated
Formulation 0.01M sodium phosphate with 0.15M NaCl and no preservative; pH 7.2
Gene Alias Horseradish Peroxidase
Host Species Goat
Immunogen Peroxidase [Horseradish]
Quantity 2 mL
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.